Learn More
Novus Biologicals™ Human IL-12 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191183
Catalog No. NBP191183
Add to cart
Description
Description
The Human IL-12 p70/IL-12A ELISA Kit (Colorimetric) measures human IL-12 p70/IL-12A in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
3.1 - 200pg/mL
3592
ELISA Kit
2.1pg/mL
Human (Hu)
IL-12
IL12A
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-12 p70, Biotin-Conjugate anti-human IL-12 p70 monoclonal antibody, Streptavidin-HRP, Human IL-12 p70 Standard lyophilized, 400 pg/ml upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Immunology, Innate Immunity
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha cha