Learn More
Novus Biologicals™ Human IL-13 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191176
Catalog No. NBP191176
Add to cart
Description
Description
The Human IL-13 ELISA Kit (Colorimetric) measures human IL-13 in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
1.6 - 100pg/mL
3596
ELISA Kit
0.7pg/mL
Human (Hu)
IL-13
IL13
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-13, Biotin-Conjugate anti-human IL-13 monoclonal antibody, Streptavidin-HRP, Human IL-13 Standard lyophilized, 200 pg/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Asthma, Cancer, Cytokine Research, Immunology
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600