Learn More
R&D Systems™ Human IL-13 Luminex Performance Assay
Bead-based multiplex assay for the Luminex™ platform
Manufacturer:  R&D Systems™ LUB213
Catalog No. LUB213
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • User selects specific analytes of interest
  • Requires small sample size
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
Specifications
Specifications
15.6pg/mL
1 Kit
38.00 - 28000pg/mL
3596
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Polystyrene
Human
IL-13
IL13
Bead-based Multiplex Assay for the Luminex™ Platform
Cell Culture Supernates (50μL),Serum (15μL),EDTA Plasma (15μL),Heparin Plasma (15μL)