Learn More
Bon Opus Biosciences
Recombinant Human Interleukin-15/IL-15
Manufacturer: Bon Opus Biosciences C0161MG
Recombinant Human Interleukin-15 is produced by our E.coli expression system and the target gene encoding Asn49-Ser162 is expressed. Bon Opus Cat. #C016