Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-18 BPa Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1191
This item is not returnable.
View return policy
Description
IL-18 BPa/IL18BP Monoclonal specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for Neutralization.Specifications
IL-18 BPa/IL18BP | |
Monoclonal | |
LYOPH | |
Neutralization 5-20 ug/mL | |
AAD17190 | |
IL18BP | |
Mouse myeloma cell line NS0-derived recombinant human IL-18 BPa Thr29-Gly192 Accession # AAD17190 | |
500 μg | |
Primary | |
Detects human IL-18 BPa in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) IL-18 BPc or rmIL-18 BPd is observed. | |
Human | |
Purified |
Neutralization | |
136007 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL18 BPa, IL18BP, IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10068 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction