Learn More
Raybiotech IncSupplier Diversity Partner Human IL-36 gamma (IL-1 F9) ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHIL1F91
For cell culture supernatants, plasma, and serum samples. Sensitivity: 13 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human IL-1F1 (IL-1 alpha), IL-1F2 (IL-1 beta), IL-1F3 (IL-1ra), IL-1F4 (IL-18), IL-1F5 (IL-36ra), IL-1F6 (IL-36 alpha), IL-1F7 (IL-37), IL-1F8 (IL-36 beta), IL-1F10 (IL-38), IL-1F11 (IL-33)
Catalog No. 50-194-9418