Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-2 ELISA Kits Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.

Interleukin 2 Recombinant protein SIL2 from Thermo Scientific™ specifically detects Interleukin 2 in samples, and it is validated for ELISA and Western Blot

$496.00 - $1838.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Target Human
View More Specs

Products 3
Catalog Number Mfr. No. Description Sufficient For Price Quantity & Availability  
Catalog Number Mfr. No. Description Sufficient For Price Quantity & Availability  
ENEH2IL2 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
EH2IL2
Human IL-2 ELISA Kit 96 microplate wells
Each for $496.00
Add to cart
 
ENEH2IL22 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
EH2IL22
Human IL-2 ELISA Kit 2 x 96 microplate wells
Each for $872.00
Add to cart
 
ENEH2IL25 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
EH2IL25
Human IL-2 ELISA Kit 5 x 96 microplate wells
Pack of 5 for $1,838.00
Add to cart
 
Description

Description

Assay IL-2 (human interleukin-2) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-2 ELISA Kits.

These human IL-2 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-2 (IL-2) in samples at 50 to 100µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-2 antibody pairs and IL-2 standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-2 (IL-2)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 38.4 to 1500pg/mL

  • Sensitivity: < 6pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50µL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, TNFβ, GM-CSF, INFγ, or mouse IL-2

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Intra-assay CV <10%; inter-assay CV <10%
Human
Absorbance at 450nm minus absorbance at 550nm
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
SDS
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit