Learn More
Raybiotech IncSupplier Diversity Partner Human IL-32 alpha ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHIL32A1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human IL-32 alpha.
Catalog No. 50-194-9448