Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-33 DuoSet ELISA

Catalog No. p-7079219
Click to view available options
Quantity:
1 Kit (for 15 Plates)
1 Kit (for 5 Plates)
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Human IL-33 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-33 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-33.

Specifications

Description Fucose Detection
Product Type ELISA Kit
Quantity 1 Kit (for 15 Plates)
Species Human
Assay Range 23.40 to 1500 pg/mL
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.