Learn More
Gibco™ Human IL-33 Recombinant Protein, PeproTech®

E. coli Recombinant Protein
$87.00 - $5616.00
Specifications
Accession Number | O95760 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 90865 |
Molecular Weight (g/mol) | 17.9 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
20033100UG
![]() ![]() |
Gibco™
20033100UG |
100 µg |
Each for $1,124.00
|
|
|||||
200-33-10UG
![]() ![]() |
Gibco™
2003310UG |
10 µg |
Each for $211.00
|
|
|||||
200-33-1MG
![]() ![]() |
Gibco™
200331MG |
1 mg |
Each for $5,616.00
|
|
|||||
20033250UG
![]() ![]() |
Gibco™
20033250UG |
250 µg |
Each for $1,966.00
|
|
|||||
200-33-2UG
![]() ![]() |
Gibco™
200332UG |
2 µg |
Each for $87.00
|
|
|||||
20033500UG
![]() ![]() |
Gibco™
20033500UG |
500 µg |
Each for $3,370.00
|
|
|||||
200-33-50UG
![]() ![]() |
Gibco™
2003350UG |
50 µg |
Each for $674.00
|
|
|||||
Description
IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer's patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.Specifications
O95760 | |
protein with no preservative | |
17.9 | |
E. coli | |
RUO | |
9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; IL1F11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
90865 | |
Human IL-33 | |
-20°C | |
<1 EU/µg | |
IL33 | |
Unconjugated | |
SITGISPITE YLASLSTYND QSITFALEDE SYEIYVEDLK KDEKKDKVLL SYYESQHPSN ESGDGVDGKM LMVTLSPTKD FWLHANNKEH SVELHKCEKP LPDQAFFVLH NMHSNCVSFE CKTDPGVFIG VKDNHLALIK VDSSENLCTE NILFKLSET | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.