Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-34 ELISA Kit

Catalog No. EH275RB
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human IL-34 quantitates human IL-34 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-34.

IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2010004A03Rik; AI593503; C16orf77; IL34; IL-34; ILN; Interleukin; interleukin 34; Interleukin34; interleukin-34
Assay Range 7 to 250 pg/mL
Assay Sensitivity 7 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 146433
Gene Symbol Il34
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL-34
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.