Learn More
Shenandoah Biotechnology, IncSupplier Diversity Partner Human IL-35, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial  DFS Item
Manufacturer:  Shenandoah Biotechnology, IncSupplier Diversity Partner 1004910UG
Human IL-35 is produced in HEK 293 cells. This recombinant protein has a purity limit of ≥95% and a low endotoxin level of ≤5.00 EUs/ug. The biological activity is measured in a Not active assay with an ED50 < N/A . This product is lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, 30 mM sodium chloride, pH7.5, and is shipped at Ice pack. When this product is recieved, reconstitute with sterile water at 0.1 mg/mL. ACF= Animal Component Free
Catalog No. 50-712-783