Learn More
Gibco™ Human IL-4 Receptor alpha (soluble) Recombinant Protein, PeproTech®

CHO cells Recombinant Protein
$87.00 - $4320.00
Specifications
Accession Number | P24394 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3566 |
Molecular Weight (g/mol) | 23.9 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
20004RC100
![]() ![]() |
Gibco™
20004RC100UG |
100 µg |
Each for $864.00
|
|
|||||
20004RC1MG
![]() ![]() |
Gibco™
20004RC1MG |
1 mg |
Each for $4,320.00
|
|
|||||
20004RC20UG
![]() ![]() |
Gibco™
20004RC20UG |
20 µg |
Each for $211.00
|
|
|||||
20004RC250
![]() ![]() |
Gibco™
20004RC250UG |
250 µg |
Each for $1,512.00
|
|
|||||
20004RC500
![]() ![]() |
Gibco™
20004RC500UG |
500 µg |
Each for $2,592.00
|
|
|||||
20004RC5UG
![]() ![]() |
Gibco™
20004RC5UG |
5 µg |
Each for $87.00
|
|
|||||
Description
IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.Specifications
P24394 | |
protein with no preservative | |
23.9 | |
CHO cells | |
RUO | |
582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4 R subunit alpha; IL4RA; IL-4 RA; IL-4 R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
3566 | |
Human IL-4 Receptor alpha (soluble) | |
-20°C | |
<1 EU/µg | |
IL4R | |
Unconjugated | |
GNMKVLQEPT CVSDYMSIST CEWKMNGPTN CSTELRLLYQ LVFLLSEAHT CIPENNGGAG CVCHLLMDDV VSADNYTLDL WAGQQLLWKG SFKPSEHVKP RAPGNLTVHT NVSDTLLLTW SNPYPPDNYL YNHLTYAVNI WSENDPADFR IYNVTYLEPS LRIAASTLKS GISYRARVRA WAQCYNTTWS EWSPSTKWHN SYREPFEQH | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.