Learn More
Gibco™ Human IL-4 Recombinant Protein, PeproTech®

E. coli Recombinant Protein
$87.00 - $3510.00
Specifications
Accession Number | P05112 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3565 |
Molecular Weight (g/mol) | 15.1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
20004100UG
![]() ![]() |
Gibco™
20004100UG |
100 µg |
Each for $702.00
|
|
|||||
200-04-1MG
![]() ![]() |
Gibco™
200041MG |
1 mg |
Each for $3,510.00
|
|
|||||
200-04-20UG
![]() ![]() |
Gibco™
2000420UG |
20 µg |
Each for $211.00
|
|
|||||
20004250UG
![]() ![]() |
Gibco™
20004250UG |
250 µg |
Each for $1,229.00
|
|
|||||
20004500UG
![]() ![]() |
Gibco™
20004500UG |
500 µg |
Each for $2,106.00
|
|
|||||
200-04-50UG
![]() ![]() |
Gibco™
2000450UG |
50 µg |
Each for $422.00
|
|
|||||
200-04-5UG
![]() ![]() |
Gibco™
200045UG |
5 µg |
Each for $87.00
|
|
|||||
Description
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
P05112 | |
protein with no preservative | |
15.1 | |
E. coli | |
RUO | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4 delta2; interleukin 4 variant IL-4 delta3; interleukin 4 variant IL-4 int2A; interleukin 4 variant IL-4 int3A; interleukin 4 variant IL-4 int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
3565 | |
Human IL-4 | |
-20°C | |
<1 EU/µg | |
Il4 | |
Unconjugated | |
MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.