Learn More
Novus Biologicals™ Human IL-5 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191274
Catalog No. NBP191274
Add to cart
Description
Description
The Human IL-5 ELISA Kit (Colorimetric) measures human IL-5 in cell culture supernatant, serum.
Specifications
Specifications
7.8 - 500pg/mL
3567
ELISA Kit
1.5pg/mL
Human (Hu)
IL-5
IL5
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-5, Biotin-Conjugate anti-human IL-5 monoclonal antibody, Streptavidin-HRP, Human IL-5 Standard lyophilized, 1ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I