Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Human IL-5 Platinum ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of human IL-5.

Manufacturer:  Invitrogen™ BMS278TEN

 View more versions of this product

Catalog No. 50-182-17


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-5
Biotin-Conjugate anti-human IL-5 monoclonal antibody
Streptavidin-HRP
Human IL-5 Standard lyophilized, 1ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Specifications

Specifications

P05113
Biotin
3567
Adhesive Plate Covers
10 x 96 tests
1.5 pg/mL
50μL
Human
1 hr 20 min
Human
B cell differentiation factor 1, TRF, EDF
6.6%
7.8-500 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Serum, Supernatants
Serum (50μL), Supernatants (50μL)
2-8°C
3 hr 10 min
IL5
6.8%
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only