Learn More
R&D Systems™ Human IL-6 High Sensitivity Luminex Performance Assay
Bead-based multiplex assay for the Luminex™ platform
Manufacturer:  R&D Systems™ LHSC206
Catalog No. LHSC206
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • User selects specific analytes of interest
  • Requires small sample size
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
Specifications
Specifications
Polystyrene
1 Kit
0.99 - 4050pg/mL
3569
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Serum, Plasma (EDTA, Heparin)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
0.31pg/mL
Human
IL-6
IL6
Bead-based Multiplex Assay for the Luminex™ Platform
Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)