Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL12RB1 Full-length ORF (AAH29121.1) Recombinant Protein without tag

Used for AP, Func, Screening

Manufacturer:  Abnova™ H00003594G01

Catalog No. 89-948-631


Add to cart

Description

Description

The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukine 12 (IL12) with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: MEPLVTWVVPLLFLFLLSRQGAACRTSECCFQDPPYPDADSGSASGPRDLRCYRISSDRYECSWQYEGPTAGVSHFLRCCLSSGRCCYFAAGSATRLQFSDQAGVSVLYTVTLWVESWARNQTEKSPEVTLQLYNSVKYEPPLGDIKVSKLAGQLRMEWETPDNQVGAEVQFRHRTPSSPWKLGDCGPQDDDTESCLCPLEMNVAQEFQLRRRRLGSQGSSWSKWSSPVCVPPENPPQPQVRFSVEQLGQDGRRRLTLKEQPTQLELPEGCQGLAPGTEVTYRLQLHMLSCPCKAKATRTLHLGKMPYLSGAAYNVAVISSNQFGPGLNQTWHIPADTHTDGMISAHCNLRLPDSRDSPASASRVAGITGICHHTRLILYF
Specifications

Specifications

AAH29121.1
Antibody Production, Compound Screening, Functional Study
Liquid
42.4kDa
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD212|IL-12R-BETA1|IL12RB|MGC34454
IL12RB1
Yes
wheat germ expression system with proprietary liposome technology
Liquid
3594
IL12RB1 (Human) Recombinant Protein
MEPLVTWVVPLLFLFLLSRQGAACRTSECCFQDPPYPDADSGSASGPRDLRCYRISSDRYECSWQYEGPTAGVSHFLRCCLSSGRCCYFAAGSATRLQFSDQAGVSVLYTVTLWVESWARNQTEKSPEVTLQLYNSVKYEPPLGDIKVSKLAGQLRMEWETPDNQVGAEVQFRHRTPSSPWKLGDCGPQDDDTESCLCPLEMNVAQEFQLRRRRLGSQGSSWSKWSSPVCVPPENPPQPQVRFSVEQLGQDGRRRLTLKEQPTQLELPEGCQGLAPGTEVTYRLQLHMLSCPCKAKATRTLHLGKMPYLSGAAYNVAVISSNQFGPGLNQTWHIPADTHTDGMISAHCNLRLPDSRDSPASASRVAGITGICHHTRLILYF
RUO
IL12RB1
Wheat Germ (in vitro)
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit