Learn More
Abnova™ Human IL12RB2 Full-length ORF (NP_001550.1, 1 a.a. - 862 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00003595P01
Description
The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense. [provided by RefSeq]
Sequence: MAHTFRGCSLAFMFIITWLLIKAKIDACKRGDVTVKPSHVILLGSTVNITCSLKPRQGCFHYSRRNKLILYKFDRRINFHHGHSLNSQVTGLPLGTTLFVCKLACINSDEIQICGAEIFVGVAPEQPQNLSCIQKGEQGTVACTWERGRDTHLYTEYTLQLSGPKNLTWQKQCKDIYCDYLDFGINLTPESPESNFTAKVTAVNSLGSSSSLPSTFTFLDIVRPLPPWDIRIKFQKASVSRCTLYWRDEGLVLLNRLRYRPSNSRLWNMVNVTKAKGRHDLLDLKPFTEYEFQISSKLHLYKGSWSDWSESLRAQTPEEEPTGMLDVWYMKRHIDYSRQQISLFWKNLSVSEARGKILHYQVTLQELTGGKAMTQNITGHTSWTTVIPRTGNWAVAVSAANSKGSSLPTRINIMNLCEAGLLAPRQVSANSEGMDNILVTWQPPRKDPSAVQEYVVEWRELHPGGDTQVPLNWLRSRPYNVSALISENIKSYICYEIRVYALSGDQGGCSSILGNSKHKAPLSGPHINAITEEKGSILISWNSIPVQEQMGCLLHYRIYWKERDSNSQPQLCEIPYRVSQNSHPINSLQPRVTYVLWMTALTAAGESSHGNEREFCLQGKANWMAFVAPSICIAIIMVGIFSTHYFQQKVFVLLAALRPQWCSREIPDPANSTCAKKYPIAEEKTQLPLDRLLIDWPTPEDPEPLVISEVLHQVTPVFRHPPCSNWPQREKGIQGHQASEKDMMHSASSPPPPRALQAESRQLVDLYKVLESRGSDPKPENPACPWTVLPAGDLPTHDGYLPSNIDDLPSHEAPLADSLEELEPQHISLSVFPSSSLHPLTFSCGDKLTLDQLKMRCDSLMLSpecifications
NP_001550.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
123.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL12RB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3595 | |
IL12RB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAHTFRGCSLAFMFIITWLLIKAKIDACKRGDVTVKPSHVILLGSTVNITCSLKPRQGCFHYSRRNKLILYKFDRRINFHHGHSLNSQVTGLPLGTTLFVCKLACINSDEIQICGAEIFVGVAPEQPQNLSCIQKGEQGTVACTWERGRDTHLYTEYTLQLSGPKNLTWQKQCKDIYCDYLDFGINLTPESPESNFTAKVTAVNSLGSSSSLPSTFTFLDIVRPLPPWDIRIKFQKASVSRCTLYWRDEGLVLLNRLRYRPSNSRLWNMVNVTKAKGRHDLLDLKPFTEYEFQISSKLHLYKGSWSDWSESLRAQTPEEEPTGMLDVWYMKRHIDYSRQQISLFWKNLSVSEARGKILHYQVTLQELTGGKAMTQNITGHTSWTTVIPRTGNWAVAVSAANSKGSSLPTRINIMNLCEAGLLAPRQVSANSEGMDNILVTWQPPRKDPSAVQEYVVEWRELHPGGDTQVPLNWLRSRPYNVSALISENIKSYICYEIRVYALSGDQGGCSSILGNSKHKAPLSGPHINAITEEKGSILISWNSIPVQEQMGCLLHYRIYWKERDSNSQPQLCEIPYRVSQNSHPINSLQPRVTYVLWMTALTAAGESSHGNEREFCLQGKANWMAFVAPSICIAIIMVGIFSTHYFQQKVFVLLAALRPQWCSREIPDPANSTCAKKYPIAEEKTQLPLDRLLIDWPTPEDPEPLVISEVLHQVTPVFRHPPCSNWPQREKGIQGHQASEKDMMHSASSPPPPRALQAESRQLVDLYKVLESRGSDPKPENPACPWTVLPAGDLPTHDGYLPSNIDDLPSHEAPLADSLEELEPQHISLSVFPSSSLHPLTFSCGDKLTLDQLKMRCDSLML | |
RUO | |
IL12RB2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.