Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL15RA Partial ORF (NP_002180, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name IL15RA (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.74kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-948-666 View Documents Abnova™
H00003601Q01L
25 ug Each for $599.28
Add to cart
 
89-948-665 View Documents Abnova™
H00003601Q01S
10 ug Each for $384.10
Add to cart
 
Description

Description

The protein encoded by this gene is a cytokine receptor that specifically binds IL15 with high affinity. The receptors of IL15 and IL2 share two subunits, the IL2R beta and IL2R gamma chains. This forms the basis of many overlapping biological activities of IL15 and IL2. The IL2 receptor requires an additional IL2-specific alpha subunit for high affinity IL2 binding. This protein is structurally related to IL2R alpha, but is capable of binding IL15 with high affinity independent of other subunits, which suggests the distinct roles between IL15 and IL2. This receptor is reported to enhance cell proliferation and expression of apoptosis inhibitor BCL2L1/BCL2-XL and BCL2. Multiple alternatively spliced transcript variants of this gene have been reported. The full length sequences of only two variants encoding distinct isoforms are available. [provided by RefSeq]

Sequence: ITCPPPMSVEHADIWVKSYSLYSRERYICNSGFKRKAGTSSLTECVLNKATNVAHWTTPSLKCIRDPALVHQRPAPPSTVTTAGVTPQPESLSPSGKEPA
Specifications

Specifications

IL15RA (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.74kDa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit