Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL16 Full-length ORF (AAH40272.1, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name IL16 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 74.8kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-948-671 View Documents Abnova™
H00003603P01L
25 ug Each for $599.28
Add to cart
 
89-948-670 View Documents Abnova™
H00003603P01S
10 ug Each for $384.10
Add to cart
 
Description

Description

The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MESHSRAGKSRKSAKFRSISRSLMLCNAKTSDDGSSPDEKYPDPFEISLAQGKEGIFHSSVQLADTSEAGPSSVPDLALASEAAQLQAAGNDRGKTCRRIFFMKESSTASSREKPGKLEAQSSNFLFPKACHQRARSNSTSVNPYCTREIDFPMTKKSAAPTDRQPYSLCSNRKSLSQQLDCPAGKAAGTSRPTRSLSTAQLVQPSGGLQASVISNIVLMKGQAKGLGFSIVGGKDSIYGPIGIYVKTIFAGGAAAADGRLQEGDEILELNGESMAGLTHQDALQKFKQAKKGLLTLTVRTRLTAPPSLCSHLSPPLCRSLSSSTCITKDSSSFALESPSAPISTAKPNYRIMVEVSLQKEAGVGLGIGLCSVPYFQCISGIFVHTLSPGSVAHLDGRLRCGDEIVEISDSPVHCLTLNEVYTILSHCDPGPVSIIVSRHPDPQKEEMEAQEVK
Specifications

Specifications

IL16 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
74.8kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit