Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human IL17C Full-length ORF (NP_037410.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00027189P01L
Description
The protein encoded by this gene is a T cell-derived cytokine that shares the sequence similarity with IL17. This cytokine was reported to stimulate the release of tumor necrosis factor alpha and interleukin 1 beta from a monocytic cell line. The expression of this cytokine was found to be restricted to activated T cells. [provided by RefSeq]
Sequence: MTLLPGLLFLTWLHTCLAHHDPSLRGHPHSHGTPHCYSAEELPLGQAPPHLLARGAKWGQALPVALVSSLEAASHRGRHERPSATTQCPVLRPEEVLEADTHQRSISPWRYRVDTDEDRYPQKLAFAECLCRGCIDARTGRETAALNSVRLLQSLLVLRRRPCSRDGSGLPTPGAFAFHTEFIHVPVGCTCVLPRSVSpecifications
NP_037410.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
RUO | |
IL17C | |
Wheat Germ (in vitro) | |
GST Tag | |
Liquid |
Protein Array, ELISA, Western Blot | |
27189 | |
Human IL17C Full-length ORF (NP_037410.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
CX2, IL-17C, IL-21, MGC126884, MGC138401 | |
IL17C | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction