Learn More
Abnova™ Human IL17F (Q96PD4) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4409
Catalog No. 89-948-695
Add to cart
Description
Description
The protein encoded by this gene is a cytokine that shares sequence similarity with IL17. This cytokine is expressed by activated T cells, and has been shown to stimulate the production of several other cytokines, including IL6, IL8, and CSF2/GM_CSF. This cytokine is also found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1. [provided by RefSeq]
Sequence: MRKIPKVGHTFFQKPESCPPVPGGSMKLDIGIINENQRVSMSRNIESRSTSPWNYTVTWDPNRYPSEVVQAQCRNLGCINAQGKEDISMNSVPIQQETLVVRRKHQGCSVSFQLEKVLVTVGCTCVTPVIHHVQ
Specifications
Specifications
Q96PD4
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
30kDa
Escherichia coli expression system
25 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
IL17F
The activity is determined by the dose-dependent production of IL-6 by mouse NIH 3T3 fibroblasts. The expected ED50 for this effect is 28-42ng/mL.
Yes
Escherichia coli expression system
Lyophilized
112744
IL17F (Human) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MRKIPKVGHTFFQKPESCPPVPGGSMKLDIGIINENQRVSMSRNIESRSTSPWNYTVTWDPNRYPSEVVQAQCRNLGCINAQGKEDISMNSVPIQQETLVVRRKHQGCSVSFQLEKVLVTVGCTCVTPVIHHVQ
RUO
IL-17F|ML-1|ML1
IL17F
Escherichia coli
None