Learn More
Abnova™ Human IL1B (P01584, 117 a.a. - 269 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3626
Catalog No. 89-948-742
Add to cart
Description
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is produced by activated macrophages as a proprotein, which is proteolytically processed to its active form by caspase 1 (CASP1/ICE). This cytokine is an important mediator of the inflammatory response, and is involved in a variety of cellular activities, including cell proliferation, differentiation, and apoptosis. The induction of cyclooxygenase-2 (PTGS2/COX2) by this cytokine in the central nervous system (CNS) is found to contribute to inflammatory pain hypersensitivity. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. [provided by RefSeq]
Sequence: APVRSLNCTLRDSQQKSLVMSGPYELKALHLQGQDMEQQVVFSMSFVQGEESNDKIPVALGLKEKNLYLSCVLKDDKPTLQLESVDPKNYPKKKMEKRFVFNKIEINNKLEFESAQFPNWYISTSQAENMPVFLGGTKGGQDITDFTMQFVSS
Specifications
Specifications
P01584
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
17kDa
Escherichia coli expression system
>90% by SDS-PAGE and HPLC
APVRSLNCTLRDSQQKSLVMSGPYELKALHLQGQDMEQQVVFSMSFVQGEESNDKIPVALGLKEKNLYLSCVLKDDKPTLQLESVDPKNYPKKKMEKRFVFNKIEINNKLEFESAQFPNWYISTSQAENMPVFLGGTKGGQDITDFTMQFVSS
RUO
IL-1|IL1-BETA|IL1F2
IL1B
Escherichia coli
None
Escherichia coli expression system
Lyophilized
3553
IL1B (Human) Recombinant Protein
Ion exchange column and HPLC reverse phase column
10 ug
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg (1 EU/μg)
IL1B
The ED50 as determined by the dose-dependent proliferation of murine D10S cells, was found to be ≤ 2.0 pg/mL, corresponding to a specific activity of ≥ 2 x 109 units/mg.
Yes