Learn More
Abnova™ Human IL1F9 (Q9NZH8) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6099
Catalog No. 89-948-764
Add to cart
Description
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. The activity of this cytokine is mediated by interleukin 1 receptor-like 2 (IL1RL2/IL1R-rp2), and is specifically inhibited by interleukin 1 family, member 5 (IL1F5/IL-1 delta). Interferon-gamma, tumor necrosis factor-alpha and interleukin 1, beta (IL1B) are reported to stimulate the expression of this cytokine in keratinocytes. The expression of this cytokine in keratinocytes can also be induced by a contact hypersensitivity reaction or herpes simplex virus infection. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. [provided by RefSeq]
Sequence: SMCKPITGTINDLNQQVWTLQGQNLVAVPRSDSVTPVTVAVITCKYPEALEQGRGDPIYLGIQNPEMCLYCEKVGEQPTLQLKEQKIMDLYGQPEPVKPFLFYRAKTGRTSTLESVAFPDWFIASSKRDQPIILTSELGKSYNTAFELNIND
Specifications
Specifications
Q9NZH8
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
17.0kDa
E. coli expression system
10 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
IL1F9
Measured by its ability to induce IL-8 production of human PBMCs.
Escherichia coli
None
E. coli expression system
Lyophilized
56300
IL1F9 (Human) Recombinant Protein
0.95
SMCKPITGTINDLNQQVWTLQGQNLVAVPRSDSVTPVTVAVITCKYPEALEQGRGDPIYLGIQNPEMCLYCEKVGEQPTLQLKEQKIMDLYGQPEPVKPFLFYRAKTGRTSTLESVAFPDWFIASSKRDQPIILTSELGKSYNTAFELNIND
RUO
IL-1F9|IL-1H1|IL-1RP2|IL1E|IL1H1|IL1RP2
IL1F9
Human ;Human
Yes