Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL1RAPL1 Partial ORF (NP_055086.1, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011141Q01S

 View more versions of this product

Catalog No. 89-948-776


Add to cart

Description

Description

The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. It is most closely related to interleukin 1 receptor accessory protein-like 2 (IL1RAPL2). This gene and IL1RAPL2 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. Deletions and mutations in this gene were found in patients with mental retardation. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities. [provided by RefSeq]

Sequence: FEKAELSKSKEISCRDIEDFLLPTREPEILWYKECRTKTWRPSIVFKRDTLLIREVREDDIGNYTCELKYGGFVVRRTTELTVTAPLTDKPPKLLYPMES
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FEKAELSKSKEISCRDIEDFLLPTREPEILWYKECRTKTWRPSIVFKRDTLLIREVREDDIGNYTCELKYGGFVVRRTTELTVTAPLTDKPPKLLYPMES
RUO
IL1RAPL1
Wheat Germ (in vitro)
GST
NP_055086.1
Liquid
11141
IL1RAPL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IL1R8/IL1RAPL/MRX10/MRX21/MRX34/OPHN4/TIGIRR-2
IL1RAPL1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit