Learn More
Abnova™ Human IL28A (Q8IZJ0, 175 amino acids) Partial Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P4174
Catalog No. 89-948-859
Add to cart
Description
Description
This gene encodes a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA). [provided by RefSeq]
Specifications
Specifications
Q8IZJ0
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
19.6kDa
Escherichia coli expression system
20 ug
RUO
IFNL2|IL-28A
IL28A
Yes
Escherichia coli expression system
Lyophilized
282616
IL28A (Human) Recombinant Protein
≥85% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
IL28A
Escherichia coli
None