Learn More
Abnova™ Human IL29 (Q8IU54, 24 a.a. - 200 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3635
Description
This gene encodes a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28A (IL28A), and interleukin 28B (IL28B) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA). [provided by RefSeq]
Sequence: TSKPTTTGKGCHIGRFKSLSPQELASFKKARDALEESLKLKNWSCSSPVFPGNWDLRLLQVRERPVALEAELALTLKVLEAAAGPALEDVLDQPLHTLHHILSQLQACIQPQPTAGPRPRGRLHHWLHRLQEAPKKESAGCLEASVTFNLFRLLTRDLKYVADGNLCLRTSTHPESTSpecifications
Q8IU54 | |
Lyophilized | |
20kDa | |
Escherichia coli expression system | |
20 ug | |
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg (1 EU/μg) | |
IL29 | |
The ED50 is determined in an anti-viral assay using human HepG2 cells infected with EMCV is typically 1-5ng/mL. | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
282618 | |
IL29 (Human) Recombinant Protein | |
Ion exchange column and HPLC reverse phase column | |
TSKPTTTGKGCHIGRFKSLSPQELASFKKARDALEESLKLKNWSCSSPVFPGNWDLRLLQVRERPVALEAELALTLKVLEAAAGPALEDVLDQPLHTLHHILSQLQACIQPQPTAGPRPRGRLHHWLHRLQEAPKKESAGCLEASVTFNLFRLLTRDLKYVADGNLCLRTSTHPEST | |
RUO | |
IFNL1/IL-29 | |
IL29 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.