Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IL3 (AAC08706, 20 aa. - 152 a.a.) Partial Recombinant Protein with His tag

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P3461

Catalog No. 89-948-873


Add to cart

Description

Description

The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMAPMTQTTSLKTSWVNCSNMIDEIITHLKQPPLPLLDFNNLNGEDQDILMENNLRRPNLEAFNRAVKSLQNASAIESILKNLLPCLPLATAAPTRHPIHIKDGDWNEFRRKLTFYLKTLENAQAQQTTLSLAIF
Specifications

Specifications

AAC08706
1 mg/mL
Liquid
3562
IL3 (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS-PAGE
MGSSHHHHHHSSGLVPRGSHMAPMTQTTSLKTSWVNCSNMIDEIITHLKQPPLPLLDFNNLNGEDQDILMENNLRRPNLEAFNRAVKSLQNASAIESILKNLLPCLPLATAAPTRHPIHIKDGDWNEFRRKLTFYLKTLENAQAQQTTLSLAIF
RUO
IL-3|MCGF|MGC79398|MGC79399|MULTI-CSF
IL3
Escherichia coli
His
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
17.3kDa
Escherichia coli expression system
>90% by SDS-PAGE
100 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
<1.0 EU/μg of protein (determined by LAL method)
IL3
The ED50 for this effect is <0.53ng/mL. Measured in a cell proliferation assay using TF1 human erythroleukemic cells.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit