Learn More
Abnova™ Human IL31 (Q6EBC2) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6095
Catalog No. 89-948-884
Add to cart
Description
Description
IL31, which is made principally by activated Th2-type T cells, interacts with a heterodimeric receptor consisting of IL31RA (MIM 609510) and OSMR (MIM 601743) that is constitutively expressed on epithelial cells and keratinocytes. IL31 may be involved in the promotion of allergic skin disorders and in regulating other allergic diseases, such as asthma (Dillon et al., 2004 [PubMed 15184896]).[supplied by OMIM]
Sequence: SHTLPVRLLRPSDDVQKIVEELQSLSKMLLKDVEEEKGVLVSQNYTLPCLSPDAQPPNNIHSPAIRAYLKTIRQLDNKSVIDEIIEHLDKLIFQDAPETNISVPTDTHECKRFILTISQQFSECMDLALKSLTSGAQQATT
Specifications
Specifications
Q6EBC2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
15.8kDa
E. coli expression system
10 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
IL31
Measured by its ability to induce STAT3 activation in human U87 cells.
Escherichia coli
None
E. coli expression system
Lyophilized
386653
IL31 (Human) Recombinant Protein
0.98
SHTLPVRLLRPSDDVQKIVEELQSLSKMLLKDVEEEKGVLVSQNYTLPCLSPDAQPPNNIHSPAIRAYLKTIRQLDNKSVIDEIIEHLDKLIFQDAPETNISVPTDTHECKRFILTISQQFSECMDLALKSLTSGAQQATT
RUO
IL-31
IL31
Human ;Human
Yes