Learn More
Abnova™ Human IL34 (Q6ZMJ4) Full-length Recombinant Protein with C-terminal His-tag expressed in HEK293 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6096
Catalog No. 89-948-899
Add to cart
Description
Description
Interleukin-34 is a cytokine that promotes the differentiation and viability of monocytes and macrophages through the colony-stimulating factor-1 receptor (CSF1R; MIM 164770) (Lin et al., 2008 [PubMed 18467591]).[supplied by OMIM]
Sequence: NEPLEMWPLTQNEECTVTGFLRDKLQYRSRLQYMKHYFPINYKISVPYEGVFRIANVTRLQRAQVSERELRYLWVLVSLSATESVQDVLLEGHPSWKYLQEVETLLLNVQQGLTDVEVSPKVESVLSLLNAPGPNLKLVRPKALLDNCFRVMELLYCSCCKQSSVLNWQDCEVPSPQSCSPEPSLQYAATQLYPPPPWSPSSPPHSTGSVRPVRAQGEGLLPHHHHHHHH
Specifications
Specifications
Q6ZMJ4
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
52.5kDa
HEK293 cells expression system
10 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
IL34
Measured by its ability to induce secretion of MCP-1 with PBMCs.
Human
His
HEK293 cells expression system
Lyophilized
146433
IL34 (Human) Recombinant Protein
0.95
NEPLEMWPLTQNEECTVTGFLRDKLQYRSRLQYMKHYFPINYKISVPYEGVFRIANVTRLQRAQVSERELRYLWVLVSLSATESVQDVLLEGHPSWKYLQEVETLLLNVQQGLTDVEVSPKVESVLSLLNAPGPNLKLVRPKALLDNCFRVMELLYCSCCKQSSVLNWQDCEVPSPQSCSPEPSLQYAATQLYPPPPWSPSSPPHSTGSVRPVRAQGEGLLPHHHHHHHH
RUO
C16orf77|IL-34|MGC34647
IL34
Human ;Human
Yes