Learn More
Abnova™ Human IL8 (P10145) (77 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P3993
Description
The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. [provided by RefSeq]
Specifications
P10145 | |
Lyophilized | |
8.4kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at 4°C for 1 week or -20°C for longer. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
IL8 | |
The ED50 was determined by its chemotaxis of hCXCR2 transfected mouse BaF/3 cells is less than 0.5ng/mL, corresponding to a specific activity of 2.0 x 106 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
3576 | |
IL8 (Human) Recombinant Protein | |
25 ug | |
RUO | |
CXCL8/GCP-1/GCP1/LECT/LUCT/LYNAP/MDNCF/MONAP/NAF/NAP-1/NAP1 | |
IL8 | |
E. coli | |
None | |
Lyophilized |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.