Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IMMP1L Full-length ORF (NP_659418.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00196294P01L

 View more versions of this product

Catalog No. 89-948-978


Add to cart

Description

Description

The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 (IMMP2L; MIM 605977) are the catalytic subunits of the IMP complex (Burri et al., 2005 [PubMed 15814844]).[supplied by OMIM]

Sequence: MLRGVLGKTFRLVGYTIQYGCIAHCAFEYVGGVVMCSGPSMEPTIQNSDIVFAENLSRHFYGIQRGDIVIAKSPSDPKSNICKRVIGLEGDKILTTSPSDFFKSHSYVPMGHVWLEGDNLQNSTDSRCYGPIPYGLIRGRIFFKIWPLSDFGFLRASPNGHRFSDD
Specifications

Specifications

NP_659418.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25059|IMP1|IMP1-LIKE
IMMP1L
Yes
wheat germ expression system
Liquid
196294
IMMP1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLRGVLGKTFRLVGYTIQYGCIAHCAFEYVGGVVMCSGPSMEPTIQNSDIVFAENLSRHFYGIQRGDIVIAKSPSDPKSNICKRVIGLEGDKILTTSPSDFFKSHSYVPMGHVWLEGDNLQNSTDSRCYGPIPYGLIRGRIFFKIWPLSDFGFLRASPNGHRFSDD
RUO
IMMP1L
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit