Learn More
Human immunodeficiency virus type 1 Goat anti-Virus, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29863
Catalog No. 89-033-708
Add to cart
Description
Description
Goat polyclonal antibody raised against human immunodeficiency virus type 1 (HIV-1).
  • High specificity
  • High reactivity
Specifications
Specifications
Human immunodeficiency virus type 1
Unconjugated
ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Liquid
Human isolate.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Virus
ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot
Goat polyclonal antibody raised against human immunodeficiency virus type 1 (HIV-1).
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Goat
IgG purification
RUO
Primary
This antibody reacts disrupted purified virions.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)