Learn More
Human immunodeficiency virus type 1 p24 Goat anti-Virus, Polyclonal, Abnova™
Goat Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29787
Catalog No. 89-033-632
Add to cart
Description
Description
Goat polyclonal antibody raised against human immunodeficiency virus type 1 p24.
  • High specificity
  • High reactivity
Specifications
Specifications
Human immunodeficiency virus type 1 p24
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Human immunodeficiency virus type 1 strain IIIB p24.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Virus
ELISA, Immunofluorescence, Western Blot
Goat polyclonal antibody raised against human immunodeficiency virus type 1 p24.
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Goat
IgG
RUO
Primary
This antibody reacts with HIV-1 p24. Does not cross-react with human T or B cells.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)