Learn More
Human immunodeficiency virus type 1 p24 Mouse anti-Virus, Clone: HM029, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13594
Catalog No. 89-033-557
Add to cart
Description
Description
Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.
  • High specificity
  • High reactivity
Specifications
Specifications
Human immunodeficiency virus type 1 p24
HM029
Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with p24 of human immunodeficiency virus type 1.
ELISA, Immunofluorescence, Lateral Flow
Unconjugated
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Liquid
IgG1
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)