Learn More
Human immunodeficiency virus type 1 p24 Mouse anti-Virus, Clone: HM082, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13596
Catalog No. 89-033-559
Add to cart
Description
Description
Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.
  • High specificity
  • High reactivity
Specifications
Specifications
Human immunodeficiency virus type 1 p24
HM082
Mouse monoclonal antibody raised against human immunodeficiency virus type 1 (HIV-1) p24.
In 10 mM PBS, pH 7.4.
Mouse
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA
Unconjugated
ELISA The optimal working dilution should be determined by the end user.
Liquid
Native purified human immunodeficiency virus type 1 antigen.
RUO
Primary
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)