Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human immunodeficiency virus type 1 p24 Rabbit anti-Virus, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB29788
Description
Rabbit polyclonal antibody raised against human immunodeficiency virus type 1 p24.
- High specificity
- High reactivity
Specifications
Human immunodeficiency virus type 1 p24 | |
Polyclonal | |
Rabbit polyclonal antibody raised against human immunodeficiency virus type 1 p24. | |
In 0.01 M Phospate buffered saline, pH 7.4 (50% glycerol). | |
Native human immunodeficiency virus antigen. | |
1 mL | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Biotin | |
ELISA The optimal working dilution should be determined by the end user. | |
Rabbit | |
Affinity purification | |
RUO | |
Virus | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction