Learn More
Novus Biologicals Human Indoleamine 2,3-dioxygenase/IDO ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262765
Description
The Human Indoleamine 2,3-dioxygenase/IDO ELISA Kit (Colorimetric) measures human Indoleamine 2,3-dioxygenase/IDO in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes Human IDO in samples. No significant cross-reactivity or interference between Human IDO and analogues was observed. | |
3620 | |
Indoleamine 2,3-dioxygenase/IDO | |
1 Kit | |
0.19 ng/mL |
0.31 - 20 ng/mL | |
ELISA | |
Storage is content dependent. | |
Human | |
Biological Fluids, Plasma, Serum | |
IDO1 |