Learn More
Raybiotech IncSupplier Diversity Partner Quantibody Human Inflammation Array 1 Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner QAHINF11
Detects 10 Human Inflammatory Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-13, IL-4, IL-6, IL-8 (CXCL8), MCP-1 (CCL2), TNF alpha
Catalog No. 50-195-0841