Learn More
Novus Biologicals™ Human iNOS ELISA Kit (Colorimetric)
SDP
This kit recognizes Human
Supplier:  Novus Biologicals™ NBP280255
Catalog No. NB013326
Add to Cart
Description
Description
No significant cross-reactivity or interference between Human NOS2/iNOS and analogues was observed.
Specifications
Specifications
P35228, P35228
46.875 pg/mL
Other Biological Fluids, Plasma, Serum
4843
Human
Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2
78.125 to 5000 pg/mL
ELISA Kit (Colorimetric)
ELISA
NOS2
1 Kit
Storage of components varies. See protocol for specific instructions.
iNOS
Product Suggestions
Product Suggestions