Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INPP5B Full-length ORF (AAH58932.1, 1 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003633P01L

 View more versions of this product

Catalog No. 89-949-078


Add to cart

Description

Description

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). This gene encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MDQSVAIQETLAEGEYCVIAVQGVLCEGDSRQSRLLGLVRYRLEHGGQEHALFLYTHRRMAITGDDVSLDQIVPVSRDFTLEEVSPDGELYILGSDVTVQLDTAELSLVFQLPFGSQTRMFLHEVARACPGFDSATRDPEFLWLSRYRCAELELEMPTPRGCNSALVTWPGYATIGGGGSNFDGLRPNGKGVPMDQSSRGQDKPESLQPRQNKSKSEITDMVRSSTITVSDKAHILSMQKFGLRDTIVKSHLLQKEEDYTYIQNFRFFAGTYNVNGQSPKECLRLWLSNGIQAPDVYCVGFQELDLSKEAFFFHDTPKEEEWFKAVSEGLHPDAKYAKVKLIRLVGIMLLLYVKQEHAAYISEVEAETVGTGIMGRMGNKGGVAIRFQFHNTSICVVNSHLAAHIEEYERRNQDYKDICSRMQFCQPDPSLPPLTISNHDVILWLGDLNYRIEELDVEKVKKLIEEKDFQMLYAYDQLKIQVAAKTVFEGFTEGELTFQPTYKYDTGSDDWDTSEKCRAPAWCDRILWKGKNITQLSYQSHMALKTSDHKPVSSVFDIGVRVVNDELYRKTLEEIVRSLDKMENANIPSVSLSKREFCFQNVKYMQLKVESFTIHNGQVPCHFEFINKPDEESYCKQWLNANPSRGFLLPDSDVEIDLELFVNKTTATKLNSGEDKIEDILVLHLDRGKDYFLSVSGNYLPSCFGSPIHTLCYMREPILDLPLETISELLAYLAAYCFETQLVTKSLI
Specifications

Specifications

AAH58932.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
111.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
5PTase|MGC65156|MGC71303
INPP5B
Yes
wheat germ expression system
Liquid
3633
INPP5B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDQSVAIQETLAEGEYCVIAVQGVLCEGDSRQSRLLGLVRYRLEHGGQEHALFLYTHRRMAITGDDVSLDQIVPVSRDFTLEEVSPDGELYILGSDVTVQLDTAELSLVFQLPFGSQTRMFLHEVARACPGFDSATRDPEFLWLSRYRCAELELEMPTPRGCNSALVTWPGYATIGGGGSNFDGLRPNGKGVPMDQSSRGQDKPESLQPRQNKSKSEITDMVRSSTITVSDKAHILSMQKFGLRDTIVKSHLLQKEEDYTYIQNFRFFAGTYNVNGQSPKECLRLWLSNGIQAPDVYCVGFQELDLSKEAFFFHDTPKEEEWFKAVSEGLHPDAKYAKVKLIRLVGIMLLLYVKQEHAAYISEVEAETVGTGIMGRMGNKGGVAIRFQFHNTSICVVNSHLAAHIEEYERRNQDYKDICSRMQFCQPDPSLPPLTISNHDVILWLGDLNYRIEELDVEKVKKLIEEKDFQMLYAYDQLKIQVAAKTVFEGFTEGELTFQPTYKYDTGSDDWDTSEKCRAPAWCDRILWKGKNITQLSYQSHMALKTSDHKPVSSVFDIGVRVVNDELYRKTLEEIVRSLDKMENANIPSVSLSKREFCFQNVKYMQLKVESFTIHNGQVPCHFEFINKPDEESYCKQWLNANPSRGFLLPDSDVEIDLELFVNKTTATKLNSGEDKIEDILVLHLDRGKDYFLSVSGNYLPSCFGSPIHTLCYMREPILDLPLETISELLAYLAAYCFETQLVTKSLI
RUO
INPP5B
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit