Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INPP5B Full-length ORF (AAH58932.1, 1 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$400.00 - $599.28

Specifications

Name INPP5B (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 111.4kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-949-078 View Documents Abnova™
H00003633P01L
25 ug Each for $599.28
Add to cart
 
89-949-077 View Documents Abnova™
H00003633P01S
10 ug Each for $400.00
Add to cart
 
Description

Description

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). This gene encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MDQSVAIQETLAEGEYCVIAVQGVLCEGDSRQSRLLGLVRYRLEHGGQEHALFLYTHRRMAITGDDVSLDQIVPVSRDFTLEEVSPDGELYILGSDVTVQLDTAELSLVFQLPFGSQTRMFLHEVARACPGFDSATRDPEFLWLSRYRCAELELEMPTPRGCNSALVTWPGYATIGGGGSNFDGLRPNGKGVPMDQSSRGQDKPESLQPRQNKSKSEITDMVRSSTITVSDKAHILSMQKFGLRDTIVKSHLLQKEEDYTYIQNFRFFAGTYNVNGQSPKECLRLWLSNGIQAPDVYCVGFQELDLSKEAFFFHDTPKEEEWFKAVSEGLHPDAKYAKVKLIRLVGIMLLLYVKQEHAAYISEVEAETVGTGIMGRMGNKGGVAIRFQFHNTSICVVNSHLAAHIEEYERRNQDYKDICSRMQFCQPDPSLPPLTISNHDVILWLGDLNYRIEELDVEKVKKLIEEKDFQMLYAYDQLKIQVAAKTVFEGFTEGELTFQPTYKYDTGSDDWDTSEKCRAPAWCDRILWKGKNITQLSYQSHMALKTSDHKPVSSVFDIGVRVVNDELYRKTLEEIVRSLDKMENANIPSVSLSKREFCFQNVKYMQLKVESFTIHNGQVPCHFEFINKPDEESYCKQWLNANPSRGFLLPDSDVEIDLELFVNKTTATKLNSGEDKIEDILVLHLDRGKDYFLSVSGNYLPSCFGSPIHTLCYMREPILDLPLETISELLAYLAAYCFETQLVTKSLI
Specifications

Specifications

INPP5B (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
111.4kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit