Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INPP5K Full-length ORF (NP_057616.2, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051763P02L

 View more versions of this product

Catalog No. 89-949-089


Add to cart

Description

Description

This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MSSRKLSGPKGRRLSIHVVTWNVASAAPPLDLSDLLQLNNRNLNLDIYVIGLQELNSGIISLLSDAAFNDSWSSFLMDVLSPLSFIKVSHVRMQGILLLVFAKYQHLPYIQILSTKSTPTGLFGYWGNKGGVNICLKLYGYYVSIINCHLPPHISNNYQRLEHFDRILEMQNCEGRDIPNILDHDLIIWFGDMNFRIEDFGLHFVRESIKNRCYGGLWEKDQLSIAKKHDPLLREFQEGRLLFPPTYKFDRNSNDYDTSEKKRKPAWTDRILWRLKRQPCAGPDTPIPPASHFSLSLRGYSSHMTYGISDHKPVSGTFDLELKPLVSAPLIVLMPEDLWTVENDMMVSYSSTSDFPSSPWDWIGLYKVGLRDVNDYVSYAWVGDSKVSCSDNLNQVYIDISNIPTTEDEFLLCYYSNSLRSVVGISRPFQIPPGSLREDPLGEAQPQI
Specifications

Specifications

NP_057616.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPS|SKIP
INPP5K
Yes
wheat germ expression system
Liquid
51763
INPP5K (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSRKLSGPKGRRLSIHVVTWNVASAAPPLDLSDLLQLNNRNLNLDIYVIGLQELNSGIISLLSDAAFNDSWSSFLMDVLSPLSFIKVSHVRMQGILLLVFAKYQHLPYIQILSTKSTPTGLFGYWGNKGGVNICLKLYGYYVSIINCHLPPHISNNYQRLEHFDRILEMQNCEGRDIPNILDHDLIIWFGDMNFRIEDFGLHFVRESIKNRCYGGLWEKDQLSIAKKHDPLLREFQEGRLLFPPTYKFDRNSNDYDTSEKKRKPAWTDRILWRLKRQPCAGPDTPIPPASHFSLSLRGYSSHMTYGISDHKPVSGTFDLELKPLVSAPLIVLMPEDLWTVENDMMVSYSSTSDFPSSPWDWIGLYKVGLRDVNDYVSYAWVGDSKVSCSDNLNQVYIDISNIPTTEDEFLLCYYSNSLRSVVGISRPFQIPPGSLREDPLGEAQPQI
RUO
INPP5K
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit