Learn More
BioworldSupplier Diversity Partner HUMAN INTERLEUKIN-3 SF9 (R DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220603841
HUMAN INTERLEUKIN-3, SF9 (RECOMBINANT): Recombinant Human Interleukin-3 glycosylated (IL-3-Sf9) Source: Insect Cells Background: Interleukin-3 (IL-3) is a cytokine produced by activated T-cells and mast cells that is active on a broa...
Catalog No. 50-253-481