Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human INTS4 Full-length ORF (NP_291025.3, 1 a.a. - 963 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00092105P01L

 View more versions of this product

Catalog No. 89-949-112


Add to cart

Description

Description

INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM]

Sequence: MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARR
Specifications

Specifications

NP_291025.3
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
134.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
INT4|MGC16733|MST093
INTS4
Yes
wheat germ expression system
Liquid
92105
INTS4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAHLKKRVYEEFTKVVQPQEEIATKKLRLTKPSKSAALHIDLCKATSPADALQYLLQFARKPVEAESVEGVVRILLEHYYKENDPSVRLKIASLLGLLSKTAGFSPDCIMDDAINILQNEKSHQVLAQLLDTLLAIGTKLPENQAIQMRLVDVACKHLTDTSHGVRNKCLQLLGNLGSLEKSVTKDAEGLAARDVQKIIGDYFSDQDPRVRTAAIKAMLQLHERGLKLHQTIYNQACKLLSDDYEQVRSAAVQLIWVVSQLYPESIVPIPSSNEEIRLVDDAFGKICHMVSDGSWVVRVQAAKLLGSMEQVSSHFLEQTLDKKLMSDLRRKRTAHERAKELYSSGEFSSGRKWGDDAPKEEVDTGAVNLIESGACGAFVHGLEDEMYEVRIAAVEALCMLAQSSPSFAEKCLDFLVDMFNDEIEEVRLQSIHTMRKISNNITLREDQLDTVLAVLEDSSRDIREALHELLCCTNVSTKEGIHLALVELLKNLTKYPTDRDSIWKCLKFLGSRHPTLVLPLVPELLSTHPFFDTAEPDMDDPAYIAVLVLIFNAAKTCPTMPALFSDHTFRHYAYLRDSLSHLVPALRLPGRKLVSSAVSPSIIPQEDPSQQFLQQSLERVYSLQHLDPQGAQELLEFTIRDLQRLGELQSELAGVADFSATYLRCQLLLIKALQEKLWNVAAPLYLKQSDLASAAAKQIMEETYKMEFMYSGVENKQVVIIHHMRLQAKALQLIVTARTTRGLDPLFGMCEKFLQEVDFFQRYFIADLPHLQDSFVDKLLDLMPRLMTSKPAEVVKILQTMLRQSAFLHLPLPEQIHKASATIIEPAGESDNPLRFTSGLVVALDVDATLEHVQDPQNTVKVQVLYPDGQAQMIHPKPADFRNPGPGRHRLITQVYLSHTAWTEACQVEVRLLLAYNSSARIPKCPWMEGGEMSPQVETSIEGTIPFSKPVKVYIMPKPARR
RUO
INTS4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit