Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IPO7 Partial ORF (NP_006382.1, 950 a.a. - 1038 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00010527Q01S

 View more versions of this product

Catalog No. 89-949-130


Add to Cart

Description

Description

The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. The protein encoded by this gene is a member of a class of approximately 20 potential Ran targets that share a sequence motif related to the Ran-binding site of importin-beta. Similar to importin-beta, this protein prevents the activation of Ran's GTPase by RanGAP1 and inhibits nucleotide exchange on RanGTP, and also binds directly to nuclear pore complexes where it competes for binding sites with importin-beta and transportin. This protein has a Ran-dependent transport cycle and it can cross the nuclear envelope rapidly and in both directions. At least four importin beta-like transport receptors, namely importin beta itself, transportin, RanBP5 and RanBP7, directly bind and import ribosomal proteins. [provided by RefSeq]

Sequence: DDEDNPVDEYQIFKAIFQTIQNRNPVWYQALTHGLNEEQRKQLQDIATLADQRRAAHESKMIEKHGGYKFSAPVVPSSFNFGGPAPGMN
Specifications

Specifications

NP_006382.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DDEDNPVDEYQIFKAIFQTIQNRNPVWYQALTHGLNEEQRKQLQDIATLADQRRAAHESKMIEKHGGYKFSAPVVPSSFNFGGPAPGMN
RUO
IPO7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10527
IPO7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14581/Imp7/MGC138673/RANBP7
IPO7
Recombinant
wheat germ expression system
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit