Learn More
Abnova™ Human IPO8 Partial ORF (NP_006381.2, 949 a.a. - 1037 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010526Q01L
Description
The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. The protein encoded by this gene is a member of a class of approximately 20 potential Ran targets that share a sequence motif related to the Ran-binding site of importin-beta. This protein binds to the nuclear pore complex and, along with RanGTP and RANBP1, inhibits the GAP stimulation of the Ran GTPase. [provided by RefSeq]
Sequence: LDLDNSVDEYQFFTQALITVQSRDAAWYQLLMAPLSEDQRTALQEVYTLAEHRRTVAEAKKKIEQQGGFTFENKGVLSAFNFGTVPSNNSpecifications
NP_006381.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LDLDNSVDEYQFFTQALITVQSRDAAWYQLLMAPLSEDQRTALQEVYTLAEHRRTVAEAKKKIEQQGGFTFENKGVLSAFNFGTVPSNN | |
RUO | |
IPO8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10526 | |
IPO8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26580/RANBP8 | |
IPO8 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.