Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IQGAP2 Partial ORF (NP_006624, 343 a.a. - 449 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010788Q01S

 View more versions of this product

Catalog No. 89-949-157


Add to cart

Description

Description

This gene encodes a member of the IQGAP family. The protein contains three IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. [provided by RefSeq]

Sequence: MYQNELFNLQKQNTMNYLAHEELLIAVEMLSAVALLNQALESNDLVSVQNQLRSPAIGLNNLDKAYVERYANTLLSVKLEVLSQGQDNLSWNEIQNCIDMVNAQIQE
Specifications

Specifications

NP_006624
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MYQNELFNLQKQNTMNYLAHEELLIAVEMLSAVALLNQALESNDLVSVQNQLRSPAIGLNNLDKAYVERYANTLLSVKLEVLSQGQDNLSWNEIQNCIDMVNAQIQE
RUO
IQGAP2
Yes
wheat germ expression system
Liquid
10788
IQGAP2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IQGAP2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit