Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRAK4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3919SP
This item is not returnable.
View return policy
Description
IRAK4 Polyclonal specifically detects IRAK4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IRAK4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
Goat | |
Affinity Purified | |
RUO | |
51135 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9NWZ3 | |
IRAK4 | |
E. coli-derived recombinant human IRAK4 Met1-Ser460 Accession # Q9NWZ3 | |
25 μg | |
Primary | |
Detects human IRAK4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) IRAK1 and rhIRAK2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction